N6-Benzyladenosine
Dodavatel: Thermo Scientific
Synonyms:
(2R,3R,4S,5R)-2-(6-(Benzylamino)-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 6-Benzylaminopurine riboside
226420050
226420010
226420050.EA
22650
CZK
226420050.
226420010.
N6-Benzyladenosine
N6-Benzyladenosine
Vzorec:
C₁₇H₁₉N₅O₄ MW: 357,36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005740 CAS číslo: 4294-16-0 EINECS: 224-298-9 |
Výsledky specifikačního testu
Appearance | white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...