Tisk…

Laminaran (from Laminaria digitata), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Laminarin

M-Clarity™ quality level = MQ200

L9634-100MG L9634-1G L9634-500MG L9634-5G
SIALL9634-100MGEA 1670 CZK
SIALL9634-100MG SIALL9634-1G SIALL9634-500MG SIALL9634-5G
Laminaran (from Laminaria digitata), Sigma-Aldrich®
Laminaran

Polysaccharide substrate for laminarinase


Laminarin from Laminaria digitata has been used:
as a stimulant to study its effect on the respiratory burst using haemocytes of P. vannamei
as a carbon source to study its effect on enzyme production using Pichia membranifaciens culture
as a carbon source for Polaribacter sp. culture

Vzorec: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00466918
CAS číslo: 9008-22-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR