Cyrene™ ≥98.5%, BioRenewable DMF and NMP substitute, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
Cyrene has been applied in replacement of NMP and/or DMF in some applications, such as demonstrating to exceed NMP′s dispersive ability for graphene solutions due to its optimum polarity and high viscosity, creating larger and less defective graphene flakes.
- 99% biodegradation in 28 days
- Stable during incineration
- Not mutagenic or genotoxic
- ASTM D6866 - Standard Test Methods for Determining the Biobased Content
- Made from Renewable Resource − Cellulose
CYRENE is a trademark of Circa Group Pvt Ltd
Vzorec:
C₆H₈O₃ Bod varu: 227 °C Hustota: 1,25 g/mL |
MDL Number:
MFCD23105760 CAS číslo: 53716-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...