Vanadyl sulphate hydrate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Vanadium (IV) sulphate oxide hydrate, Vanadium(IV) oxide sulphate hydrate, Vanadium oxide sulphate hydrate, Basic vanadium (IV) sulphate hydrate
M-Clarity™ quality level = MQ100
233706-25G
233706-100G
233706-500G
SIAL233706-25GEA
2780
CZK
SIAL233706-25G
SIAL233706-100G
SIAL233706-500G
Vanadyl sulphate hydrate, Sigma-Aldrich®
Vanadyl sulphate hydrate
Vzorec:
O₅SV·H₂O |
MDL Number:
MFCD00149933 CAS číslo: 123334-20-3 EINECS: 248-652-7 UN: 2931 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...