Tisk…
5 mg, tablets, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate
M-Clarity™ quality level = MQ300
N9389-100TAB N9389-50TAB
SIALN9389-100TABEA 11590 CZK
SIALN9389-100TAB SIALN9389-50TAB
5 mg, tablets, Sigma-Aldrich®
4-Nitrofenylfosforečnan dvojsodný hexahydrát (pNPP)
Chromogenic substrate for the determination of acid and alkaline phosphatases.
Vzorec: C₆H₄NNa₂O₆P·6H₂O
MW: 371.15
Bod tání: 300
Storage Temperature: Freezer
MDL Number: MFCD00066288
CAS číslo: 333338-18-4
EINECS: 224-246-5