Tisk…
≥97%, powder, BioReagent cell culture reagent, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate
M-Clarity™ quality level = MQ200
N4645-1G N4645-5G
SIALN4645-1GEA 6190 CZK
SIALN4645-1G SIALN4645-5G
≥97%, powder, BioReagent cell culture reagent, Sigma-Aldrich®
4-Nitrofenylfosforečnan dvojsodný hexahydrát (pNPP)
Chromogenic substrate for the determination of acid and alkaline phosphatases.
Vzorec: C₆H₄NNa₂O₆P·6H₂O
MW: 371.15
Bod tání: 300
Storage Temperature: Freezer
MDL Number: MFCD00066288
CAS číslo: 333338-18-4
EINECS: 224-246-5