Tisk…

Magnesium aluminate ≥99% (metals basis)

Dodavatel: Thermo Fisher Scientific

Synonyms: Dialuminium magnesium tetraoxide, Magnesium aluminum oxide, Aluminum magnesium oxide

22950.22 22950.36 22950.PG
22950.22EA 1720 CZK
22950.22 22950.36 22950.PG
Magnesium aluminate ≥99% (metals basis)
Magnesium aluminate

Purity: 99% CAS No.: 12068-51-8 Molecular Formula: MgAl2O4 Formula Weight: 142.26 Physical Form: -325 Mesh Powder Odour: Odourless MDL No.: MFCD00049475

Vzorec: Al₂MgO₄
MW: 142.27
Storage Temperature: Ambient
MDL Number: MFCD00049475
CAS číslo: 12068-51-8
EINECS: 235-100-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR