Dodecyl-β-D-glucopyranoside, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
D8035-1G
D8035-5G
SIALD8035-1GEA
10780
CZK
SIALD8035-1G
SIALD8035-5G
Dodecyl-β-D-glucopyranoside, Sigma-Aldrich®
Dodecyl-β-D-glucopyranoside
Vzorec:
C₁₈H₃₆O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00063298 CAS číslo: 59122-55-3 EINECS: 261-614-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...