Tisk…

Dichloroisocyanuric acid sodium salt dihydrate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: NaDCC

35915-1KG 35915-250G 35915-50G
SIAL35915-1KGEA 0 CZK
SIAL35915-1KG SIAL35915-250G SIAL35915-50G
Dichloroisocyanuric acid sodium salt dihydrate, Sigma-Aldrich®
Dichloroisocyanuric acid sodium salt dihydrate
Vzorec: C₃Cl₂N₃NaO₃·2H₂O
MW: 255.98
Storage Temperature: Ambient
MDL Number: MFCD00149404
CAS číslo: 51580-86-0
EINECS: 220-767-7
UN: 3077
ADR: 9 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR