Tisk…

Diisopropylcarbodiimide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC

M-Clarity™ quality level = MQ200

38370-100ML 38370-25ML 38370-500ML
SIAL38370-100MLEA 10040 CZK
SIAL38370-100ML SIAL38370-25ML SIAL38370-500ML
Diisopropylcarbodiimide, Sigma-Aldrich®
Diisopropylcarbodiimide
Vzorec: C₇H₁₄N₂
MW: 126,2 g/mol
Bod varu: 145 °C (1013 hPa)
Hustota: 0,81 g/cm³ (20 °C)
Bod vzplanutí: 33 °C
MDL Number: MFCD00065689
CAS číslo: 693-13-0
EINECS: 211-743-7
UN: 2929
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR