Di-tert-butylcarbodiimide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,3-Di-tert-butylcarbodiimide, N,N'-Di-tert-butylcarbodiimide, N,N-di-tert-butylcarbodiimide, N,N'-methanetetraylbis(1,1-dimethylethylamine)
M-Clarity™ quality level = MQ200
235563-1G
235563-5G
SIAL235563-1GEA
2220
CZK
SIAL235563-1G
SIAL235563-5G
Di-tert-butylcarbodiimide, Sigma-Aldrich®
Di-tert-butylcarbodiimide
Vzorec:
C₉H₁₈N₂ MW: 154,26 g/mol |
MDL Number:
MFCD00010231 CAS číslo: 691-24-7 EINECS: 211-719-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...