Tisk…
Dextran, M.W. ~ 70,000 (from Leuconostoc sp.), BioChemika®, Sigma-Aldrich®
Dodavatel: Merck
31390-100G 31390-25G 31390-500G
SIAL31390-100GEA 15440 CZK
SIAL31390-100G SIAL31390-25G SIAL31390-500G
Dextran, M.W. ~ 70,000 (from Leuconostoc sp.), BioChemika®, Sigma-Aldrich®
Dextran
Vzorec: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS číslo: 9004-54-0
EINECS: 232-677-5