Calcium aluminate ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
22646.06
22646.14
22646.22
22646.06EA
2470
CZK
22646.06
22646.14
22646.22
Calcium aluminate ≥99% (metals basis)
Calcium aluminate
Purity: 99%
CAS No.: 12042-68-1
Molecular Formula: CaAl2O4
Formula Weight: 158.02
Physical Form: -200 Mesh Powder
Density: 2.981
Melting Point: 1600°
MDL No.: MFCD00049722
Vzorec:
Al₂CaO₄ MW: 158.04 Bod tání: 1600 °C Hustota: 2.981 Storage Temperature: Ambient |
MDL Number:
MFCD00049722 CAS číslo: 12042-68-1 EINECS: 234-931-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...