Tisk…

Eskulin seskihydrát, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6,7-Dihydroxycoumarin 6-glucoside sesquihydrate, Esculin sesquihydrate

M-Clarity™ quality level = MQ200

02350-10G 02350-50G
SIAL02350-10GEA 3720 CZK
SIAL02350-10G SIAL02350-50G
Eskulin seskihydrát, Sigma-Aldrich®
Eskulin seskihydrát
Vzorec: C₁₅H₁₆O₉·1,5H₂O
MW: 367.31
Bod tání: 203…205 °C
MDL Number: MFCD00149492
CAS číslo: 66778-17-4
EINECS: 208-517-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR