Eskulin seskihydrát ≥97%
Dodavatel: Thermo Scientific
Synonyms:
6,7-Dihydroxycoumarin 6-glucoside sesquihydrate, Esculin sesquihydrate
A11624.06
A11624.14
A11624.22
A11624.06EA
1190
CZK
A11624.06
A11624.14
A11624.22
Eskulin seskihydrát ≥97%
Eskulin seskihydrát
Purity: 97%
CAS No.: 66778-17-4
Molecular Formula: C15H16O9.1.5H2O
Formula Weight: 367.31 (340.28anhy)
Physical Form: Powder
Sensitivity: Air Sensitive and Hygroscopic
Appearance: White
Melting Point: 203-205°
Beilstein Reference: 95387
Merck Reference: 14,3698
Optical Rotation: -41° (c=5 in pyridine)
MDL No.: MFCD00149492
Inhibits chemically induced carcinogenesis in mouse skin and kidney
Vzorec:
C₁₅H₁₆O₉·1,5H₂O MW: 367,31 g/mol Bod tání: 203…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149492 CAS číslo: 66778-17-4 EINECS: 208-517-5 Merck Index: 14,3698 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...