5-Amino-2-naphthalenesulphonic acid 97%
Dodavatel: Thermo Scientific
Synonyms:
5-Amino-2-naphthalenesulphonic Acid, 5-Amino-β-naphthalenesulphonic acid, 1-Naphthylamine-6-sulfonic acid, 1,6-Cleve's acid, 5-Aminonaphthalene-2-sulphonic acid, 5-Aminonaphthalene-2-sulfonic acid
L03099.06
L03099.14
L03099.06EA
951
CZK
L03099.06
L03099.14
5-Amino-2-naphthalenesulphonic acid 97%
5-Amino-2-naphthalenesulphonic acid
Purity: 97%
CAS No.: 119-79-9
Molecular Formula: C10H9NO3S
Formula Weight: 223.25
Odour: Odourless
Melting Point:
Vzorec:
C₁₀H₉NO₃S MW: 223,25 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004030 CAS číslo: 119-79-9 EINECS: 204-351-2 Merck Index: 14,2350 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...