4-Fluoro-1-naphthoic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
A16778.03
A16778.06
A16778.03EA
1820
CZK
A16778.03
A16778.06
4-Fluoro-1-naphthoic acid ≥97%
4-Fluoro-1-naphthoic acid
Purity: 97%
CAS No.: 573-03-5
Molecular Formula: C11H7FO2
Formula Weight: 190.18
Melting Point: 223-226°
Beilstein Reference: 2257292
Vzorec:
C₁₁H₇FO₂ MW: 190,17 g/mol Bod tání: 223…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004011 CAS číslo: 573-03-5 EINECS: 209-346-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...