Tisk…

Kyselina 4-amino-3-hydroxy-1-naftalensulfonová 99%

Dodavatel: Thermo Fisher Scientific
Varování

B24339.14 B24339.22
B24339.14EA 2540 CZK
B24339.14 B24339.22
Kyselina 4-amino-3-hydroxy-1-naftalensulfonová 99%
Kyselina 4-amino-3-hydroxy-1-naftalensulfonová

Purity: 99% CAS No.: 116-63-2 Molecular Formula: C10H9NO4S Formula Weight: 239.25 Melting Point: ca 295° dec. Beilstein Reference: 2697469 Merck Reference: 14,454 MDL No.: MFCD00004019

Vzorec: C₁₀H₉NO₄S
Bod varu: 466 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00004019
CAS číslo: 116-63-2
EINECS: 204-147-3
Merck Index: 14,454

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR