Kyselina 4-amino-3-hydroxy-1-naftalensulfonová ≥90% ACS
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Amino-2-naphthol-4-sulphonic acid, 4-Amino-3-hydroxy-α-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulphonic acid, 1-Amino-2-hydroxy-4-naphthalenesulphonic acid
Purity: 90%
CAS No.: 116-63-2
Molecular Formula: C10H9NO4S
Formula Weight: 239.25
Physical Form: Powder
Melting Point: ca 295° dec.
Beilstein Reference: 2697469
Merck Reference: 14,454
MDL No.: MFCD00004019
Vzorec:
C₁₀H₉NO₄S MW: 239,25 g/mol Bod varu: 466 °C (1013 hPa) Bod tání: ∼ 295 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00004019 CAS číslo: 116-63-2 EINECS: 204-147-3 Merck Index: 14,454 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...