Tisk…

Kyselina chromtropová dvojsodná sůl dihydrát ACS

Dodavatel: Thermo Scientific
Varování

Synonyms: 4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate

22414.06 22414.14 22414.22
22414.06EA 1060 CZK
22414.06 22414.14 22414.22
Kyselina chromtropová dvojsodná sůl dihydrát ACS
Kyselina chromtropová dvojsodná sůl dihydrát

CAS No.: 5808-22-0 Molecular Formula: C10H6Na2O8S2.2H2O Formula Weight: 400.29 (364.26anhy) Physical Form: Powder Appearance: Off-White to Buff-Coloured Melting Point: 300° Beilstein Reference: 3647189 Merck Reference: 14,2241 MDL No.: MFCD00150612

Vzorec: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400,29 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150612
CAS číslo: 5808-22-0
EINECS: 204-972-9
Merck Index: 14,2241

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR