Tisk…

2-Naphthoyl chloride ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Naphthoyl Chloride

A14046.09 A14046.18 A14046.30
A14046.09EA 1930 CZK
A14046.09 A14046.18 A14046.30
2-Naphthoyl chloride ≥98%
2-Naphthoyl chloride

Purity: 98% CAS No.: 2243-83-6 Molecular Formula: C11H7ClO Formula Weight: 190.63 Flash Point: 110°(230°F) Odour: Pungent Boiling Point: 159-160°/9mm Beilstein Reference: 907776 UN No.: 3261 MDL No.: MFCD00004093

Vzorec: C₁₁H₇ClO
MW: 190,63 g/mol
Bod varu: 159…160 °C (9 mmHg)
Bod tání: 48…52 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004093
CAS číslo: 2243-83-6
EINECS: 218-822-5
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR