Tisk…

N,N-Dimethyl-p-fenylendiamoniumsulfát 99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N,N-Dimethyl-1,4-phenylenediammonium sulphate, N,N-Dimethylbenzene-1,4-diammonium sulphate

181430250 181431000
ACRO181430250EA 2660 CZK
ACRO181430250 ACRO181431000
N,N-Dimethyl-p-fenylendiamoniumsulfát 99%
N,N-Dimethyl-p-fenylendiamoniumsulfát
Vzorec: C₈H₁₄N₂O₄S
MW: 234,28 g/mol
Bod tání: 234…236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012992
CAS číslo: 536-47-0
EINECS: 208-636-2
UN: 2811
ADR: 6.1,II

Objednat

Výsledky specifikačního testu

Appearance White to grey to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR