Tisk…
AlphaTec® 87-118 Natural Rubber Latex Gloves
Dodavatel: Ansell
87118/9.5 87118/10.5 87118/6.5 87118/7.5 87118/8.5
ANSE87118/9.5EA 1500 CZK
ANSE87118/9.5 ANSE87118/10.5 ANSE87118/6.5 ANSE87118/7.5 ANSE87118/8.5
AlphaTec® 87-118 Natural Rubber Latex Gloves
Rukavice
Thick natural rubber latex gloves, offering heavy-duty protection and chemical resistance.
  • Easy donning and doffing, and limited liquid roll-off
  • Diamond grip pattern - Heightened grip performance
These protective gloves also feature a beaded cuff, which facilitates donning and doffing, while preventing droplets travelling beyond the glove’s surface, for greater wrist protection.
Designed with a diamond grip pattern, these chemical-resistant gloves also offer wearers secure grip, for more confident handling.
Composed of flexible natural latex rubber, these AlphaTec® gloves also boast 100% cotton flock lining, which absorbs some sweat and minimises discomfort.
Recommended for loading and unloading of process equipment, loading raw material, transferring liquids and solids between vessels and tanks and process equipment, assembly and disassembly of equipment, production line support and maintenance.
Application: Automotive aftermarket, chemical, machinery and equipment, metal fabrication
Certifikace: EN ISO 21420:2020.