Tisk…

1,4-Dibutylbenzene ≥97%

Dodavatel: Thermo Scientific

L16396.03 L16396.09
L16396.03EA 1340 CZK
L16396.03 L16396.09
1,4-Dibutylbenzene ≥97%
1,4-Dibutylbenzene

Purity: 97+% CAS No.: 1571-86-4 Molecular Formula: C14H22 Formula Weight: 190.33 Flash Point: 110°(230°F) Refractive Index: 1.4890 Melting Point: -31° Boiling Point: 227° Beilstein Reference: 1858055 MDL No.: MFCD01076591

Vzorec: C₁₄H₂₂
MW: 190,33 g/mol
Bod varu: 227 °C
Bod tání: –31 °C
Hustota: 0,856
Bod vzplanutí: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01076591
CAS číslo: 1571-86-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR