Tisk…

ABT-491 hydrochloride ≥98% (by HPLC), Sigma-Aldrich®

Dodavatel: Merck

A9227-25MG A9227-5MG
SIALA9227-25MGEA 28620 CZK
SIALA9227-25MG SIALA9227-5MG
ABT-491 hydrochloride ≥98% (by HPLC), Sigma-Aldrich®
ABT-491 hydrochloride

Highly potent, selective and orally active platelet-activating factor (PAF) receptor antagonist. Ki = 0.6 nM in human platelets.

Vzorec: C₂₈H₂₂FN₅O₂·xHCl
Storage Temperature: Refrigerator
MDL Number: MFCD00945301
CAS číslo: 189689-94-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR