Tisk…

CD437 ≥98% (by HPLC), solid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

C5865-25MG C5865-5MG
SIALC5865-25MGEA 19750 CZK
SIALC5865-25MG SIALC5865-5MG
CD437 ≥98% (by HPLC), solid, Sigma-Aldrich®
CD437

CD437 is a retinoic acid receptor (RAR)γ-selective agonist, γ-selective retinoid; potent inducer of apoptosis.

Vzorec: C₂₇H₂₆O₃
Storage Temperature: Freezer
MDL Number: MFCD03106506
CAS číslo: 125316-60-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR