ReadiPrep™ PEG Virus Precipitation Kit
Dodavatel: AAT BIOQUEST
Designed for consistent and dependable results. The ReadiPrep™ PEG virus precipitation kit provides a streamlined approach for the concentration of viral particles and the elimination of contaminants, circumventing the need for ultracentrifugation.
- The use of non-toxic reagents in the procedure ensures safe handling
- Can concentrate a diverse array of viruses, including retroviruses, baculoviruses, lentiviruses, and phages, from cell culture media and environmental samples, with a concentration increase of over 100-fold
- Six-month shelf life
This kit is well-suited for both small-scale laboratory samples with low viral titers and large-scale virus preparations with high yields and titers.
Informace o dodávce: Shipped with blue ice.
Upozornení: Research use only.