Tisk…
Cryogenic Labels for Inkjet Printers (A4 Size)
Dodavatel: GA
A4CIJS-23WH A4CIJS-32WH A4CIJS-8WH
GAINA4CIJS-23WHEA 2350 CZK
GAINA4CIJS-23WH GAINA4CIJS-32WH GAINA4CIJS-8WH
Cryogenic Labels for Inkjet Printers (A4 Size)
Štítky
Cryogenic inkjet labels, provided in A4 sheet format, for printing with most standard desktop inkjet printers. Perfect for identifying cryo vials, tubes, bags, and boxes destined for cryogenic storage in liquid nitrogen tanks (–196 °C) and lab freezers (–80 °C).
  • Temperature range: –196 to +121 °C
  • Resist extreme cryogenic and deep-freeze temperatures in a liquid phase and vapor phase Liquid Nitrogen
  • Print eye-catching color logos, graphics, images, and data
  • Resists wiping with ethanol and surface sanitising wipes
  • Requires zero cure time; can be immersed in liquid nitrogen directly following application
These inkjet printable cryogenic labels are ideal for identifying cryo vials, tubes, plastic bags, as well as cardboard and polycarbonate freezer boxes destinated for cryo storage in liquid nitrogen (–196 °C) and lab freezers (–80 °C) with eye-catching color logos, graphics, images, and data.
The obtained printout offers an impressive resistance to harsh chemicals, such as alcohol, acetone, and xylene, as well as to wiping and rubbing with alcohol sanitising wipes.