Anilin ≥99.0%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
|
Nebezpečí
|
Synonyms:
Aminobenzene, Phenylamine
21424.230EA
465
CZK
21424.230
21424.290
Anilin ≥99.0%, GPR RECTAPUR®
Anilin
* material may darken during storage with no loss in purity
Vzorec:
C₆H₅NH₂ Bod varu: 184 °C (1013 hPa) Bod tání: –6,2 °C Hustota: 1,019 g/cm³ (20 °C) Bod vzplanutí: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007629 CAS číslo: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1,II REACH: 01-2119451454-41 Merck Index: 13,00661 |
Výsledky specifikačního testu
Appearance | colorless to dark-brownish / yellow liquid |
Assay (GC) | min. 99.0% |
Water | max. 0.2% |
Ash content | max. 0.02% |
Density (20°C) | 1.020 - 1.025 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...