Tisk…
alkynyl-dT-Phosphoramidite
Dodavatel: AAT BIOQUEST
6214 6215
AATB6214EA 7100 CZK
AATB6214 AATB6215
alkynyl-dT-Phosphoramidite
alkynyl-dT-Phosphoramidite
Alkynyl-dT-Phosphoramidite is an excellent building block for introducing an alkyne group into an oligo via dT.
  • Stable for 6 months
Quality tested in-house