Tisk…
Anti-MeCP2 Mouse Monoclonal Antibody [clone: 4F11]
Dodavatel: ANTIBODIES.COM
A104324-100 A104324-50
ANTIA104324-100EA 14820 CZK
ANTIA104324-100 ANTIA104324-50
Anti-MeCP2 Mouse Monoclonal Antibody [clone: 4F11]
Protilátky
Mouse monoclonal (4F11) antibody to MeCP2 for WB, ICC/IF and IHC with samples derived from human, rat, mouse, horse, canine, bovine and porcine.
  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:5,000 to 1:10,000, ICC/IF: 1:1,000 to 1:5,000, IHC: 1:1,000 to 1:5,000
Type: Primary
Antigen: MeCP2
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse, Horse, Dog, Bovine, Porcine