Tisk…

Anti-MeCP2 Chicken Polyclonal Antibody

Dodavatel: ANTIBODIES.COM
A104322-100 A104322-50
ANTIA104322-100EA 14820 CZK
ANTIA104322-100 ANTIA104322-50
Anti-MeCP2 Chicken Polyclonal Antibody
Protilátky
Chicken polyclonal antibody to MeCP2 for WB, ICC/IF and IHC with samples derived from human, monkey, rat and mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:10,000 to 1:20,000, ICC/IF: 1:1,000 to 1:2,000, IHC: 1:1,000 to 1:2,000

Type: Primary
Antigen: MeCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Monkey, Rat, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR