Tisk…

Anti-BCMA Rabbit Recombinant Antibody [clone: DM6] (Biotin)

Dodavatel: ANTIBODIES.COM
A317437-100 A317437-500
ANTIA317437-100EA 26370 CZK
ANTIA317437-100 ANTIA317437-500
Anti-BCMA Rabbit Recombinant Antibody [clone: DM6] (Biotin)
Protilátky
Recombinant rabbit monoclonal [DM6] antibody to BCMA (Biotin) for ELISA and Flow Cytometry with samples derived from human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: BCMA
Clonality: Recombinant
Clone: DM6
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR