Tisk…
Anti-BCMA Rabbit Recombinant Antibody [clone: DM16] (Biotin)
Dodavatel: ANTIBODIES.COM
A317429-100 A317429-500
ANTIA317429-100EA 26370 CZK
ANTIA317429-100 ANTIA317429-500
Anti-BCMA Rabbit Recombinant Antibody [clone: DM16] (Biotin)
Protilátky
Recombinant rabbit monoclonal [DM16] antibody to BCMA (Biotin) for ELISA and Flow Cytometry with samples derived from human.
  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: 1:100, IP: 1:30
Type: Primary
Antigen: BCMA
Clonality: Recombinant
Clone: DM16
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human