Tisk…
Anti-ACE2 Rabbit Recombinant Antibody [clone: DM47] (Biotin)
Dodavatel: ANTIBODIES.COM
A317417-100 A317417-500
ANTIA317417-100EA 26370 CZK
ANTIA317417-100 ANTIA317417-500
Anti-ACE2 Rabbit Recombinant Antibody [clone: DM47] (Biotin)
Protilátky
Recombinant rabbit monoclonal [DM47] antibody to ACE2 (Biotin) for ELISA and Flow Cytometry with samples derived from human.
  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100
Type: Primary
Antigen: ACE2
Clonality: Recombinant
Clone: DM47
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human