Tisk…

Anti-ACE2 Rabbit Recombinant Antibody [clone: DM48] (Biotin)

Dodavatel: ANTIBODIES.COM
A317416-100 A317416-500
ANTIA317416-100EA 26370 CZK
ANTIA317416-100 ANTIA317416-500
Anti-ACE2 Rabbit Recombinant Antibody [clone: DM48] (Biotin)
Protilátky
Recombinant rabbit monoclonal [DM48] antibody to ACE2 (Biotin) for ELISA and Flow Cytometry with samples derived from human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: ACE2
Clonality: Recombinant
Clone: DM48
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR