Tisk…

Anti-MeCP2 Rabbit Polyclonal Antibody

Dodavatel: ANTIBODIES.COM
A85427-100 A85427-50
ANTIA85427-100EA 14820 CZK
ANTIA85427-100 ANTIA85427-50
Anti-MeCP2 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody to MeCP2 for WB, ICC/IF and IHC with samples derived from Human, Rat, Mouse and Monkey.

  • Validated applications: WB, ICC/IF, IHC
  • Recommended dilutions: WB: 1:10000 to 1:20000, ICC/IF: 1:2000, IHC: 1:200

Type: Primary
Antigen: MeCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse;Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR