Tisk…

Anti-Lamin A + Lamin C Mouse Monoclonal Antibody [clone: 4C4]

Dodavatel: ANTIBODIES.COM
A85444-100 A85444-50
ANTIA85444-100EA 14820 CZK
ANTIA85444-100 ANTIA85444-50
Anti-Lamin A + Lamin C Mouse Monoclonal Antibody [clone: 4C4]
Protilátky
Mouse monoclonal (4C4) antibody to Lamin A + Lamin C for WB and ICC/IF with samples derived from Human.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:1000 to 1:2000, ICC/IF: 1:100

Type: Primary
Antigen: Lamin A + Lamin C
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR