Tisk…
Anti-Lamin A + Lamin C Chicken Polyclonal Antibody
Dodavatel: ANTIBODIES.COM
A85443-100 A85443-50
ANTIA85443-100EA 14820 CZK
ANTIA85443-100 ANTIA85443-50
Anti-Lamin A + Lamin C Chicken Polyclonal Antibody
Protilátky
Chicken polyclonal antibody to Lamin A + Lamin C for WB and ICC/IF with samples derived from Human, Rat, Mouse, Horse, Monkey and Canine.
  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:2000, ICC/IF: 1:100
Type: Primary
Antigen: Lamin A + Lamin C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human;Rat;Mouse;Horse;Monkey;Dog