Tisk…

Anti-procollagen type IIA Mouse Monoclonal Antibody [clone: ABT-CII]

Dodavatel: ANTIBODIES.COM
A96501-100 A96501-50
ANTIA96501-100EA 13540 CZK
ANTIA96501-100 ANTIA96501-50
Anti-procollagen type IIA Mouse Monoclonal Antibody [clone: ABT-CII]
Protilátky
Mouse monoclonal (ABT-CII) antibody to Collagen Type II for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: procollagen type IIA
Clonality: Monoclonal
Clone: ABT-CII
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR