Tisk…

Anti-Progesterone Receptor Mouse Monoclonal Antibody [clone: ABT-PR]

Dodavatel: ANTIBODIES.COM
A95519-100 A95519-50
ANTIA95519-100EA 13540 CZK
ANTIA95519-100 ANTIA95519-50
Anti-Progesterone Receptor Mouse Monoclonal Antibody [clone: ABT-PR]
Protilátky
Mouse monoclonal (ABT-PR) antibody to Progesterone Receptor for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Progesterone Receptor
Clonality: Monoclonal
Clone: ABT-PR
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR