Tisk…
Anti-OX40L/TNFSF4 Rabbit Recombinant Antibody [clone: DM112]
Dodavatel: ANTIBODIES.COM
A318595-100 A318595-500
ANTIA318595-100EA 18240 CZK
ANTIA318595-100 ANTIA318595-500
Anti-OX40L/TNFSF4 Rabbit Recombinant Antibody [clone: DM112]
Protilátky
Recombinant rabbit monoclonal [DM112] antibody to OX40L/TNFSF4 for ELISA and flow cytometry with samples derived from human.
  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: OX40L/TNFSF4
Clonality: Recombinant
Clone: DM112
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human