Tisk…
Anti-Interferon alpha 2 Rat Monoclonal Antibody [clone: KT107]
Dodavatel: ANTIBODIES.COM
A124-100 A124-200
ANTIA124-100EA 12690 CZK
ANTIA124-100 ANTIA124-200
Anti-Interferon alpha 2 Rat Monoclonal Antibody [clone: KT107]
Protilátky
Rat monoclonal (KT107) antibody to IFNA2 for ELISA with samples derived from human.
  • Validated applications: ELISA
  • Recommended dilutions: ELISA: 1/1000, WB: 1/500 to 1/10000
Type: Primary
Antigen: Interferon alpha 2
Clonality: Monoclonal
Clone: KT107
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human