Tisk…

Anti-SERT Rabbit Polyclonal Antibody

Dodavatel: ANTIBODIES.COM
A104336-100 A104336-50
ANTIA104336-100EA 14820 CZK
ANTIA104336-100 ANTIA104336-50
Anti-SERT Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody to SERT for WB and ICC/IF with samples derived from human, Bovine, Rat and mouse.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: ICC/IF: 1:2000 to 1:5000

Type: Primary
Antigen: SERT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Bovine;Rat;Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR