Tisk…

Anti-Luciferase Goat Polyclonal Antibody

Dodavatel: ANTIBODIES.COM
A121637-15 A121637-600
ANTIA121637-15EA 18100 CZK
ANTIA121637-15 ANTIA121637-600
Anti-Luciferase Goat Polyclonal Antibody
Protilátky
Goat polyclonal antibody to Luciferase for WB with samples derived from Photinus pyralis.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2,000, IF: 1:25 to 1:250

Type: Primary
Antigen: Luciferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Photinus pyralis
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR