Tisk…

Anti-Eph Receptor A4/SEK Recombinant Recombinant Antibody [clone: DMC472]

Dodavatel: ANTIBODIES.COM
A318718-100 A318718-500
ANTIA318718-100EA 18240 CZK
ANTIA318718-100 ANTIA318718-500
Anti-Eph Receptor A4/SEK Recombinant Recombinant Antibody [clone: DMC472]
Protilátky
Recombinant chimeric monoclonal [DMC472] antibody to Eph Receptor A4/SEK for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: Eph Receptor A4/SEK
Clonality: Recombinant
Clone: DMC472
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR