Tisk…
Anti-cTnI Mouse Monoclonal Antibody [clone: KT143]
Dodavatel: ANTIBODIES.COM
A166-100 A166-200
ANTIA166-100EA 12690 CZK
ANTIA166-100 ANTIA166-200
Anti-cTnI Mouse Monoclonal Antibody [clone: KT143]
Protilátky
Mouse monoclonal (KT143) antibody to cTnI for WB, ELISA and sELISA with samples derived from human.
  • Validated applications: WB, ELISA, sELISA
  • Recommended dilutions: ELISA: 1 µg/ml, WB: 1/500 to 1/5000, sELISA: 1 µg/ml
Type: Primary
Antigen: cTnI
Clonality: Monoclonal
Clone: KT143
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human