Tisk…
Anti-AFP Mouse Monoclonal Antibody [clone: KT137]
Dodavatel: ANTIBODIES.COM
A147-100 A147-200
ANTIA147-100EA 12260 CZK
ANTIA147-100 ANTIA147-200
Anti-AFP Mouse Monoclonal Antibody [clone: KT137]
Protilátky
Mouse monoclonal (KT137) antibody to AFP for sELISA with samples derived from human.
  • Validated applications: sELISA
  • Recommended dilutions: sELISA: 3 µg/ml
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: KT137
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human