Tisk…

Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (APC (Allophycocyanin)/iFluor® 700)

Dodavatel: AAT BIOQUEST
50198 50199
AATB50198EA 7870 CZK
AATB50198 AATB50199
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (APC (Allophycocyanin)/iFluor® 700)
Protilátky
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Upozornení: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)/iFluor® 700
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR