Tisk…
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 568)
Dodavatel: AAT BIOQUEST
50082 50083
AATB50082EA 4090 CZK
AATB50082 AATB50083
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 568)
Protilátky
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.
  • Affinity purified
  • Easily reconstituted
Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).
Upozornení: For research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 568
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human